Skip to Content
MilliporeSigma
All Photos(1)

Documents

750352

Sigma-Aldrich

3-(Fmoc-amino)benzonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16N2O2
Molecular Weight:
340.37
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

165-170 °C

functional group

Fmoc

SMILES string

O=C(Nc1cccc(c1)C#N)OCC2c3ccccc3-c4ccccc24

InChI

1S/C22H16N2O2/c23-13-15-6-5-7-16(12-15)24-22(25)26-14-21-19-10-3-1-8-17(19)18-9-2-4-11-20(18)21/h1-12,21H,14H2,(H,24,25)

InChI key

SMYGNDZAWDSFSD-UHFFFAOYSA-N

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service