Skip to Content
MilliporeSigma
All Photos(1)

Documents

749605

Sigma-Aldrich

2-(Diisopropylsilyl)pyridine

95%

Synonym(s):

PyDipSiH, Pyridyldiisopropylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NSi
CAS Number:
Molecular Weight:
193.36
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

95%

refractive index

n20/D 1.496

density

0.917 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)[SiH](C(C)C)c1ccccn1

InChI

1S/C11H19NSi/c1-9(2)13(10(3)4)11-7-5-6-8-12-11/h5-10,13H,1-4H3

InChI key

XPUAXJBYLPPFKA-UHFFFAOYSA-N

Application

Reagent commonly used in acyloxylation reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.9 °F

Flash Point(C)

84.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalia Chernyak et al.
Journal of the American Chemical Society, 132(24), 8270-8272 (2010-06-01)
A new general and easily installable silicon-tethered pyridyl-containing directing group (PyDipSi) that allows for highly efficient and regioselective Pd-catalyzed ortho C-H acyloxylation of arenes has been developed. It has also been demonstrated that this directing group can efficiently be removed
The Pyridyldiisopropylsilyl Group: A Masked Functionality and Directing Group for Monoselective ortho-Acyloxylation and ortho-Halogenation Reactions of Arenes
Huang, C.; et al.
Advanced Synthesis & Catalysis, 353, 1285-1305 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service