Skip to Content
MilliporeSigma
All Photos(1)

Documents

74328

Sigma-Aldrich

1-[2-(Dimethylamino)ethyl]piperazine

≥98.0% (GC)

Synonym(s):

N,N-Dimethyl-1-piperazineethanamine, N,N-Dimethyl-2-piperazinoethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H19N3
CAS Number:
Molecular Weight:
157.26
Beilstein:
105841
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.0% (GC)

form

liquid

impurities

≤0.5% water

refractive index

n20/D 1.479

SMILES string

CN(C)CCN1CCNCC1

InChI

1S/C8H19N3/c1-10(2)7-8-11-5-3-9-4-6-11/h9H,3-8H2,1-2H3

InChI key

LGDNSGSJKBIVFG-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service