Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

735876

Sigma-Aldrich

1-(2-aminoethyl)-3-pyrrolidinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O
CAS Number:
Molecular Weight:
130.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.502

density

1.066 at 25 °C

storage temp.

2-8°C

SMILES string

NCCN1CCC(O)C1

InChI

1S/C6H14N2O/c7-2-4-8-3-1-6(9)5-8/h6,9H,1-5,7H2

InChI key

ZNLCTCNYKCENHP-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service