Skip to Content
MilliporeSigma
All Photos(1)

Documents

734721

Sigma-Aldrich

2-Iodopyridine-4-carboxylic acid

95%

Synonym(s):

2-Iodoisonicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4INO2
CAS Number:
Molecular Weight:
249.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

225-230 °C

SMILES string

OC(=O)c1ccnc(I)c1

InChI

1S/C6H4INO2/c7-5-3-4(6(9)10)1-2-8-5/h1-3H,(H,9,10)

InChI key

FALCXGCLOUKBSP-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service