Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

727342

Sigma-Aldrich

(S)-2-Aminononane

ChiPros®, produced by BASF, 99%

Synonym(s):

(S)-2-Nonylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H21N
CAS Number:
Molecular Weight:
143.27
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

produced by BASF

Assay

≥98.5% (GC)
99%

form

liquid

optical purity

enantiomeric excess: ≥98.5%

SMILES string

CCCCCCC[C@H](C)N

InChI

1S/C9H21N/c1-3-4-5-6-7-8-9(2)10/h9H,3-8,10H2,1-2H3/t9-/m0/s1

InChI key

ALXIFCUEJWCQQL-VIFPVBQESA-N

Legal Information

ChiPros is a registered trademark of BASF SE

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

165.2 °F

Flash Point(C)

74 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service