Skip to Content
MilliporeSigma
All Photos(1)

Documents

721840

Sigma-Aldrich

3,5-Dibromopyridine-4-carboxylic acid

95%

Synonym(s):

3,5-Dibromoisonicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Br2NO2
CAS Number:
Molecular Weight:
280.90
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

218-223 °C

SMILES string

OC(=O)c1c(Br)cncc1Br

InChI

1S/C6H3Br2NO2/c7-3-1-9-2-4(8)5(3)6(10)11/h1-2H,(H,10,11)

InChI key

LRKLXFGPVUZEDK-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service