Skip to Content
MilliporeSigma
All Photos(1)

Documents

720984

Sigma-Aldrich

Chloroform solution

NMR reference standard, 0.3% in acetone-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.12

grade

NMR reference standard

form

solution

concentration

0.3% in acetone-d6 (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

1H line shape

Quantity

5 mm O.D. tube contains 0.700 mL.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matteo Serino et al.
PloS one, 6(6), e21184-e21184 (2011-06-24)
Daily variations in lipid concentrations in both gut lumen and blood are detected by specific sensors located in the gastrointestinal tract and in specialized central areas. Deregulation of the lipid sensors could be partly involved in the dysfunction of glucose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service