Skip to Content
MilliporeSigma
All Photos(1)

Documents

706744

Sigma-Aldrich

1-Allyl-2-methylbenzene

97%

Synonym(s):

2-Allyltoluene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12
CAS Number:
Molecular Weight:
132.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.517

density

0.895 g/mL at 25 °C

SMILES string

Cc1ccccc1CC=C

InChI

1S/C10H12/c1-3-6-10-8-5-4-7-9(10)2/h3-5,7-8H,1,6H2,2H3

InChI key

SVIHJJUMPAUQNO-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

129.9 °F

Flash Point(C)

54.4 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service