Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

685739

Sigma-Aldrich

1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxid hexafluorophosphate, ChemDose tablets

Loading: 0.02mmol per tablet

Synonym(s):

1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxid hexafluorophosphate impregnated tablets, N-[(Dimethylamino)-1H-1,2,3-triazolo-[4,5,b]pyridin-1-ylmethylene]-N-methylmethanaminium hexafluorophosphate N-oxide impregnated tablets, ChemDose 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxid hexafluorophosphate tablets, HATU impregnated tablets

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15F6N6OP
Molecular Weight:
380.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

tablet

matrix

magnesium aluminometasilicate base material

storage temp.

2-8°C

SMILES string

CN(C)[C+](N(C)C)N1N=[N+]([O-])C2=C1C=CC=N2.F[P-](F)(F)(F)(F)F

InChI

1S/C10H15N6O.F6P/c1-13(2)10(14(3)4)15-8-6-5-7-11-9(8)16(17)12-15;1-7(2,3,4,5)6/h5-7H,1-4H3;/q+1;-1

InChI key

FKBFHOSFPRWJNV-UHFFFAOYSA-N

Legal Information

ChemDose is a trademark of Reaxa Ltd.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan J Cameron et al.
Organic & biomolecular chemistry, 17(16), 3902-3913 (2019-04-04)
The total synthesis and structural confirmation of naturally occurring all l-cyclic tetrapeptide pseudoxylallemycin A is reported. X-ray crystallography revealed that the linear precursor adopted an all-trans (ttt) extended linear conformation, while its cyclic derivative adopts a trans,cis,trans,cis (tctc) conformation. Two
Jing Zhu et al.
International journal of molecular sciences, 21(4) (2020-02-23)
: Human α-calcitonin gene-related peptide (h-α-CGRP) is a highly potent vasodilator peptide that belongs to the family of calcitonin peptides. There are two forms of CGRP receptors in humans and rodents: α-CGRP receptor predominately found in the cardiovascular system and
Hiroshi Miyazaki et al.
International journal of molecular sciences, 21(21) (2020-11-12)
The biological process of skin sensitization depends on the ability of a sensitizer to modify endogenous proteins. A direct peptide reactivity assay (DPRA), based on the biological process of skin sensitization, was developed as an alternative to controversial animal experiments.
Vincent Diemer et al.
Nature communications, 11(1), 2558-2558 (2020-05-24)
The control of cysteine reactivity is of paramount importance for the synthesis of proteins using the native chemical ligation (NCL) reaction. We report that this goal can be achieved in a traceless manner during ligation by appending a simple N-selenoethyl
J H Rothman et al.
Cancer gene therapy, 24(9), 401-408 (2017-09-25)
Peptide nucleic acid (PNA) oligomers are DNA mimics, which are capable of binding gene sequences 1000-fold more avidly than complementary native DNA by strand invasion and effectively obstruct transcription. Irreversibly obstructing the transcription or replication of a gene sequence, such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service