Skip to Content
MilliporeSigma
All Photos(1)

Documents

685372

Sigma-Aldrich

2-Amino-4,5,6,7-tetrahydrobenzothiazole

97%

Synonym(s):

4,5,6,7-Tetrahydrobenzothiazoyl-2-ylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2S
CAS Number:
Molecular Weight:
154.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

86-90 °C

SMILES string

Nc1nc2CCCCc2s1

InChI

1S/C7H10N2S/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H2,(H2,8,9)

InChI key

SMWAOXCEPHEGFV-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcin Woźnica et al.
Journal of separation science, 42(7), 1412-1422 (2019-01-27)
The objective of this article was to design the selective molecularly imprinted sorbent dedicated to the solid-phase extraction of S-pramipexole from the complex matrix such as human urine. For that purpose, S-2,6-diamino-4,5,6,7-tetrahydrobenzothiazole was used as the template acting as the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service