Skip to Content
MilliporeSigma
All Photos(1)

Documents

682640

Sigma-Aldrich

Nanoclay, surface modified

contains 25-30 wt. % methyl dihydroxyethyl hydrogenated tallow ammonium

Synonym(s):

Nanomer® I.34MN, Nanomer® clay, Montmorillonite clay

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352100

form

powder

contains

25-30 wt. % methyl dihydroxyethyl hydrogenated tallow ammonium

matrix

Montmorillonite clay base material

particle size

≤20 μm

Looking for similar products? Visit Product Comparison Guide

Application

For representative application data and nanocomposite processing methods see datasheet N605.
Recommended for melt compounding with Nylon 6, 66 and other polyamides
  • Improve modulus
  • Increase barrier
  • Enhance chemical resistance
  • Increase heat deflection temperature (HDT)

Legal Information

Nanomer® clays are products of Nanocor Corporation
Product of Nanocor®, Inc.
Nanomer is a registered trademark of Amcol International Corporation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service