Skip to Content
MilliporeSigma
All Photos(1)

Documents

680583

Sigma-Aldrich

5-Isopropyl-2-methoxyphenylboronic acid

≥95%

Synonym(s):

5-Isopropyl-2-methoxybenzeneboronic acid, [2-Methoxy-5-(1-methylethyl)phenyl]boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15BO3
CAS Number:
Molecular Weight:
194.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

mp

84-88 °C

storage temp.

2-8°C

SMILES string

COc1ccc(cc1B(O)O)C(C)C

InChI

1S/C10H15BO3/c1-7(2)8-4-5-10(14-3)9(6-8)11(12)13/h4-7,12-13H,1-3H3

InChI key

UTKAEAGLVJFBMK-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride

Application

Reactant involved in the synthesis of many biologically active molecules including:
  • Oxazolidinones for use as cholesteryl ester transfer protein inhibitors for atherosclerosis treatment
  • Selective quinazolinyl-phenol inhibitors of CHK1 as antitumor and radioprotectants
  • Biaryl-containing carbamates as CETP inhibitors
  • α -Sulfone hydroxamates as MMP inhibitors

Reactant involved in Suzuki-Miyaura cross coupling with 2-nitroarenediazonium tetrafluoroborate for synthesis of 2-nitrobiphenyls

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service