Skip to Content
MilliporeSigma
All Photos(1)

Documents

677035

Sigma-Aldrich

(2S,5S)-(+)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane

kanata purity

Synonym(s):

S,S-Me-RajPhos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21O2P
CAS Number:
Molecular Weight:
264.30
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

optical activity

[α]/D +35°, c = 1 in chloroform

refractive index

n20/D 1.5713

density

1.103 g/mL at 25 °C

SMILES string

C[C@H](P1C2=C(C=CC=C2)C3OCCO3)CC[C@@H]1C

InChI

1S/C15H21O2P/c1-11-7-8-12(2)18(11)14-6-4-3-5-13(14)15-16-9-10-17-15/h3-6,11-12,15H,7-10H2,1-2H3/t11-,12-/m0/s1

InChI key

JAKXTLMNIHPLEU-RYUDHWBXSA-N

Legal Information

Sold in collaboration with Kanata Chemical Technologies Inc. for research purposes only. These compounds were made and sold under license from E.I. du Pont de Nemours and Company, which license does not include the right to use the compounds in producing products for sale in the pharmaceutical field.
RajPhos is a trademark of Kanata Chemical Technologies, Inc.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service