Skip to Content
MilliporeSigma
All Photos(1)

Documents

671975

Sigma-Aldrich

4-Chloromethyl-5-methyl-1,3-dioxol-2-one

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5ClO3
CAS Number:
Molecular Weight:
148.54
Beilstein:
4798926
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.5% (HPLC)
97%

SMILES string

CC1=C(CCl)OC(=O)O1

InChI

1S/C5H5ClO3/c1-3-4(2-6)9-5(7)8-3/h2H2,1H3

InChI key

QCLFSYYUWPUWQR-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

264.2 °F

Flash Point(C)

129 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Iwona Dams et al.
Molecules (Basel, Switzerland), 20(12), 21346-21363 (2015-12-04)
During the process development for multigram-scale synthesis of olmesartan medoxomil (OM), two principal regioisomeric process-related impurities were observed along with the final active pharmaceutical ingredient (API). The impurities were identified as N-1- and N-2-(5-methyl-2-oxo-1,3-dioxolen-4-yl)methyl derivatives of OM. Both compounds, of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service