Skip to Content
MilliporeSigma
All Photos(1)

Documents

671517

Sigma-Aldrich

N-Fmoc-N″-succinyl-4,7,10-trioxa-1,13-tridecanediamine

95% (HPLC)

Synonym(s):

17-Oxo-6,9,12-trioxa-2,16-diazaeicosanedioic acid 1-(9H-fluoren-9-ylmethyl)ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H38N2O8
CAS Number:
Molecular Weight:
542.62
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.26

Assay

95% (HPLC)

form

solid

reaction suitability

reagent type: linker

functional group

Fmoc
amine
carboxylic acid

storage temp.

−20°C

SMILES string

O=C(CCC(O)=O)NCCCOCCCNC(OCC1C2=C(C=CC=C2)C3=C1C=CC=C3)=O

InChI

1S/C29H38N2O8/c32-27(11-12-28(33)34)30-13-5-15-36-17-19-38-20-18-37-16-6-14-31-29(35)39-21-26-24-9-3-1-7-22(24)23-8-2-4-10-25(23)26/h1-4,7-10,26H,5-6,11-21H2,(H,30,32)(H,31,35)(H,33,34)

InChI key

VWGYNWSOHMVYGG-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polyethylene glycol (PEG) reagents offer favorable characteristics for various applications, including biocompatibility and solubility.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service