Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

665738

Sigma-Aldrich

4-Pentylphenyl 4-propylbenzoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O2
CAS Number:
Molecular Weight:
310.43
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid crystal

refractive index

n20/D 1.543

density

0.989 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCCCCc1ccc(OC(=O)c2ccc(CCC)cc2)cc1

InChI

1S/C21H26O2/c1-3-5-6-8-18-11-15-20(16-12-18)23-21(22)19-13-9-17(7-4-2)10-14-19/h9-16H,3-8H2,1-2H3

InChI key

WNBFPAKRCJNBBS-UHFFFAOYSA-N

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service