Skip to Content
MilliporeSigma
All Photos(1)

Documents

653918

Sigma-Aldrich

2-Iodofluorene

98%

Synonym(s):

2-Iodo-9H-fluorene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9I
CAS Number:
Molecular Weight:
292.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

126-129 °C

SMILES string

Ic1ccc-2c(Cc3ccccc-23)c1

InChI

1S/C13H9I/c14-11-5-6-13-10(8-11)7-9-3-1-2-4-12(9)13/h1-6,8H,7H2

InChI key

VNYQUOAQPXGXQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Precursor for Organic electroluminescent devices.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry of Materials, 15, 699-699 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service