Skip to Content
MilliporeSigma
All Photos(3)

Documents

651699

Sigma-Aldrich

1-(1-Naphthylmethyl)piperazine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N2
CAS Number:
Molecular Weight:
226.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

63-67 °C (lit.)

SMILES string

C1CN(CCN1)Cc2cccc3ccccc23

InChI

1S/C15H18N2/c1-2-7-15-13(4-1)5-3-6-14(15)12-17-10-8-16-9-11-17/h1-7,16H,8-12H2

InChI key

HGYDREHWXXUUIS-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

João Anes et al.
Frontiers in microbiology, 10, 92-92 (2019-03-01)
Antimicrobial efflux is one of the important mechanisms causing multi-drug resistance (MDR) in bacteria. Chemosensitizers like 1-(1-naphthylmethyl)-piperazine (NMP) can inhibit an efflux pump and therefore can overcome MDR. However, secondary effects of NMP other than efflux pump inhibition are rarely
Sabine Schuster et al.
International journal of antimicrobial agents, 50(3), 477-481 (2017-07-12)
Drug efflux by resistance-nodulation-cell division (RND)-type transporters, such as AcrAB-TolC of Escherichia coli, is an important resistance mechanism in Gram-negative bacteria; however, its contribution to multidrug resistance (MDR) in clinical isolates is poorly defined. We inactivated acrB of a sequence
Ram Prosad Chakrabarty et al.
AMB Express, 6(1), 126-126 (2016-12-23)
The study aims at revealing the comprehensive contribution of target alteration, target protection and efflux pump to the development of high level of ciprofloxacin (CIP) resistance in Enterobacteriaceae bacteria of environmental, clinical and poultry origins. Antibiotic susceptibility test was used
Jun-Ren Sun et al.
PloS one, 7(11), e49534-e49534 (2012-11-21)
Over-expression of AdeABC efflux pump stimulated continuously by the mutated AdeRS two component system has been found to result in antimicrobial resistance, even tigecycline (TGC) resistance, in multidrug-resistant Acinetobacter baumannii (MRAB). Although the insertion sequence, ISAba1, contributes to one of
Zachary P Howard et al.
Infection and immunity, 88(12) (2020-09-16)
Coxiella burnetii is a zoonotic bacterial obligate intracellular parasite and the cause of query (Q) fever. During natural infection of female animals, C. burnetii shows tropism for the placenta and is associated with late-term abortion, at which time the pathogen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service