Skip to Content
MilliporeSigma
All Photos(1)

Documents

603953

Sigma-Aldrich

Glycolic acid-1-13C

≥99 atom % 13C, ≥99% (CP)

Synonym(s):

Hydroxyacetic acid-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH213CO2H
CAS Number:
Molecular Weight:
77.04
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Assay

≥99% (CP)

form

solid

mass shift

M+1

SMILES string

OC[13C](O)=O

InChI

1S/C2H4O3/c3-1-2(4)5/h3H,1H2,(H,4,5)/i2+1

InChI key

AEMRFAOFKBGASW-VQEHIDDOSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service