Skip to Content
MilliporeSigma
All Photos(1)

Documents

595519

Sigma-Aldrich

(S)-(−)-2,2′-Dimethoxy-1,1′-binaphthalene

97%

Synonym(s):

(S)-(−)-2,2′-Dimethoxy-1,1′-binaphthyl, (S)-1,1′-Bi-2-naphthyl dimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18O2
CAS Number:
Molecular Weight:
314.38
Beilstein:
4704232
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]20/D -52°, c = 1% in chloroform

mp

227-231 °C (lit.)

SMILES string

COc1ccc2ccccc2c1-c3c(OC)ccc4ccccc34

InChI

1S/C22H18O2/c1-23-19-13-11-15-7-3-5-9-17(15)21(19)22-18-10-6-4-8-16(18)12-14-20(22)24-2/h3-14H,1-2H3

InChI key

BJAADAKPADTRCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service