Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

591149

Sigma-Aldrich

(3,5-Dimethoxyphenyl)acetonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH2CN
CAS Number:
Molecular Weight:
177.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

54-57 °C (lit.)

SMILES string

COc1cc(CC#N)cc(OC)c1

InChI

1S/C10H11NO2/c1-12-9-5-8(3-4-11)6-10(7-9)13-2/h5-7H,3H2,1-2H3

InChI key

UUNRWZQWCNTSCV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kavya S Keremane et al.
Photochemistry and photobiology, 97(2), 289-300 (2020-10-02)
Developing effective and low-cost organic hole-transporting materials (HTMs) is crucial for the construction of high-performance perovskite solar cells (PSCs) and to promote their production in commercial ventures. In this context, we herein report the molecular design, synthesis and characterization of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service