Skip to Content
MilliporeSigma
All Photos(1)

Documents

589101

Sigma-Aldrich

Ethyl 2-ethylacrylate

96%

Synonym(s):

Ethyl α-ethylacrylate, Ethyl 2-methylenebutanoate, Ethyl ethacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(CH2)CO2C2H5
CAS Number:
Molecular Weight:
128.17
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

refractive index

n20/D 1.421 (lit.)

bp

139-145 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]\C([H])=C(\CC)C(=O)OCC

InChI

1S/C7H12O2/c1-4-6(3)7(8)9-5-2/h3-5H2,1-2H3

InChI key

OUGJKAQEYOUGKG-UHFFFAOYSA-N

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.0 °F - closed cup

Flash Point(C)

31.67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service