Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

588849

Sigma-Aldrich

trans-4-(tert-Butyldimethylsiloxy)-1-buten-1-ylboronic acid pinacol ester

95%

Synonym(s):

trans-2-[4-(tert-butyldimethylsilyloxy)-1-buten-1-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H33BO3Si
CAS Number:
Molecular Weight:
312.33
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

95%

refractive index

n20/D 1.4470 (lit.)

bp

297-298 °C (lit.)

density

0.888 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)[Si](C)(C)OCC\C=C\B1OC(C)(C)C(C)(C)O1

InChI

1S/C16H33BO3Si/c1-14(2,3)21(8,9)18-13-11-10-12-17-19-15(4,5)16(6,7)20-17/h10,12H,11,13H2,1-9H3/b12-10+

InChI key

ODTSJDLTXNDTBB-ZRDIBKRKSA-N

Application

trans-4-(tert-Butyldimethylsiloxy)-1-buten-1-ylboronic acid pinacol ester can be used as a substrate:
  • In the catalyst-free Zweifel olefination of alkenyl boronic esters using triorganocerium reagents.
  • In the study of carbosulfenylation reactions of alkenylboronates.
  • To prepare chiral tris(boronates), which are important building blocks for preparing synthetically challenging compounds.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantiomerically enriched tris (boronates): readily accessible conjunctive reagents for asymmetric synthesis
Coombs JR, et al.
Journal of the American Chemical Society, 136(46), 16140-16143 (2014)
Enantioselective, Lewis Base-Catalyzed Carbosulfenylation of Alkenylboronates by 1, 2-Boronate Migration
Tao Z, et al.
Journal of the American Chemical Society, 140(46), 15621-15625 (2018)
Catalyst-Free Enantiospecific Olefination with In Situ Generated Organocerium Species
Music A, et al.
Angewandte Chemie (International ed. in English), 58(4), 1188-1192 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service