Skip to Content
MilliporeSigma
All Photos(1)

Documents

571539

Sigma-Aldrich

[9,9-Bis(2-ethylhexyl)-9H-fluorene-2,7-diyl]bisboronic acid

≥95%

Synonym(s):

9,9-Bis(2-ethylhexyl)fluoren-2,7-diyl boronic acid, 9,9-Di(2′-ethylhexyl)fluorene-2,7-diboronic acid, 9,9-Di(2-ethylhexyl)fluorene-2,7-bisboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H44B2O4
CAS Number:
Molecular Weight:
478.28
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥95%

mp

>250 °C (lit.)

SMILES string

CCCCC(CC)CC1(CC(CC)CCCC)c2cc(ccc2-c3ccc(cc13)B(O)O)B(O)O

InChI

1S/C29H44B2O4/c1-5-9-11-21(7-3)19-29(20-22(8-4)12-10-6-2)27-17-23(30(32)33)13-15-25(27)26-16-14-24(31(34)35)18-28(26)29/h13-18,21-22,32-35H,5-12,19-20H2,1-4H3

InChI key

RBFWXVHGJQDSGG-UHFFFAOYSA-N

Application

Reactant for:
  • Coupling reactions

Other Notes

Contains varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service