Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56187

Sigma-Aldrich

(+)-Ethyl (R)-3-hydroxy-3-phenylpropionate

≥99.0% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14O3
CAS Number:
Molecular Weight:
194.23
Beilstein:
3200514
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (sum of enantiomers, GC)

optical activity

[α]20/D +51±2°, c = 1% in chloroform

refractive index

n20/D 1.510

SMILES string

CCOC(=O)C[C@@H](O)c1ccccc1

InChI

1S/C11H14O3/c1-2-14-11(13)8-10(12)9-6-4-3-5-7-9/h3-7,10,12H,2,8H2,1H3/t10-/m1/s1

InChI key

DVIBDQWVFHDBOP-SNVBAGLBSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Chiral building block; e.g. ester-amidation and reduction to an amine; Electrophilic α-amination to an α-hydrazino ester

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C. Greck et al.
Synlett, 475-475 (1993)
A. Kumar et al.
Tetrahedron Letters, 32, 1901-1901 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service