Skip to Content
MilliporeSigma
All Photos(2)

Documents

542725

Sigma-Aldrich

3-Fluorobenzenesulfonamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4SO2NH2
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

124.5-128.5 °C (lit.)

SMILES string

NS(=O)(=O)c1cccc(F)c1

InChI

1S/C6H6FNO2S/c7-5-2-1-3-6(4-5)11(8,9)10/h1-4H,(H2,8,9,10)

InChI key

CRINBBOGNYCAOV-UHFFFAOYSA-N

General description

3-Fluorobenzenesulfonamide can be synthesized by reacting 3-fluorobenzenesulfonyl chloride (in tetrahydrofuran) with 50% NH4OH.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L B Dugad et al.
Biochemistry, 28(9), 3955-3960 (1989-05-02)
Fluorine NMR has been used to examine complexes formed by 2-fluoro-, 3-fluoro-, and 2,5-difluorobenzenesulfonamide and human carbonic anhydrases I and II. The results show that all three inhibitors form complexes with both isozymes that have 2:1 inhibitor/enzyme stoichiometry. The fluorine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service