Skip to Content
MilliporeSigma
All Photos(2)

Documents

529443

Sigma-Aldrich

Cobalt(II) oxide

99.99% trace metals basis

Synonym(s):

Cobaltous oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoO
CAS Number:
Molecular Weight:
74.93
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.99% trace metals basis

form

powder

application(s)

battery manufacturing

SMILES string

O=[Co]

InChI

1S/Co.O

InChI key

IVMYJDGYRUAWML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cobalt(II) oxide (CoO) can be used as a catalytic material in the form of nanorods in oxygen-evolution reaction (OER) and photocatalytic hydrogen evolution (PHE).

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Repr. 1B - Resp. Sens. 1B - Skin Sens. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Engineering surface atomic structure of single-crystal cobalt (II) oxide nanorods for superior electrocatalysis
Ling T, et al.
Nature Communications, 7(1), 1-8 (2016)
Singlet oxygen-mediated selective C--H bond hydroperoxidation of ethereal hydrocarbons
Sagadevan A, et al.
Nature Communications, 8(1), 1-8 (2017)

Articles

Magnetic nanoparticles have attracted tremendous attention due to their novel properties and their potential applications in magnetic recording, magnetic energy storage and biomedicine.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

Magnetic materials find diverse applications from data storage to renewable energy.

Ultrasonic spray pyrolysis produces scalable nanomaterials like metal oxides and quantum dots for diverse applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service