Skip to Content
MilliporeSigma
All Photos(1)

Documents

527432

Sigma-Aldrich

tert-Butyl cinnamate

98%

Synonym(s):

trans-tert-Butyl cinnamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCOOC(CH3)3
CAS Number:
Molecular Weight:
204.26
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

bp

85 °C/0.12 mmHg (lit.)

density

1 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)OC(=O)\C=C\c1ccccc1

InChI

1S/C13H16O2/c1-13(2,3)15-12(14)10-9-11-7-5-4-6-8-11/h4-10H,1-3H3/b10-9+

InChI key

AGKLVMVJXDFIGC-MDZDMXLPSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service