Skip to Content
MilliporeSigma
All Photos(1)

Documents

525464

Sigma-Aldrich

2-Hydroxyethyl methacrylate

98%

Synonym(s):

1,2-Ethanediol mono(2-methylpropenoate), Glycol methacrylate, HEMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COOCH2CH2OH
CAS Number:
Molecular Weight:
130.14
Beilstein:
1071583
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor density

5 (vs air)

vapor pressure

0.01 mmHg ( 25 °C)

Assay

98%

contains

200 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.453 (lit.)

bp

67 °C/3.5 mmHg (lit.)

density

1.073 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OCCO

InChI

1S/C6H10O3/c1-5(2)6(8)9-4-3-7/h7H,1,3-4H2,2H3

InChI key

WOBHKFSMXKNTIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Duncan Chege et al.
Journal of acquired immune deficiency syndromes (1999), 65(5), 517-525 (2013-11-22)
To evaluate if systemic murine malarial infection enhances HIV susceptibility through parasite-induced mucosal immune alterations at sites of HIV sexual exposure. Malaria and HIV have a high degree of geographical overlap and interact substantially within coinfected individuals. We used a
Brendan P Purcell et al.
Biomaterials, 33(31), 7849-7857 (2012-07-28)
Poor cell engraftment in the myocardium is a limiting factor towards the use of bone marrow derived cells (BMCs) to treat myocardial infarction (MI). In order to enhance the engraftment of circulating BMCs in the myocardium following MI, we have
Nina Yu Kostina et al.
Biomacromolecules, 13(12), 4164-4170 (2012-11-20)
Five poly(betaine) brushes were prepared, and their resistance to blood plasma fouling was studied. Two carboxybetaines monomers were copolymerized with 2-hydroxyethyl methacrylate (HEMA) to prepare novel hydrogels. By increasing the content of the zwitterionic comonomer, a 4-fold increase in the
Y Yoshida et al.
Journal of dental research, 91(11), 1060-1065 (2012-09-13)
Previous research showed that the functional monomer 10-methacryloxydecyl dihydrogen phosphate (MDP) ionically bonds to hydroxyapatite (HAp) and forms a nano-layered structure at the interface with HAp-based substrates. Such hydrophobic nano-layering is considered to contribute to the long-term durability of the
Jie Yin et al.
Advanced materials (Deerfield Beach, Fla.), 24(40), 5441-5446 (2012-08-24)
Ordered herringbone patterns with deterministic long and short wavelengths are created using a sequential wrinkling strategy (SWS). Patterns with a prescribed zig-zag turning angle less than 90° are obtained upon sequential wrinkling of non-equi-biaxial prestrain for the first time. SWS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service