Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

525359

Sigma-Aldrich

4-(Trifluoromethoxy)phenyl isothiocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4NCS
CAS Number:
Molecular Weight:
219.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.541 (lit.)

bp

215-218 °C (lit.)

density

1.35 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)(F)Oc1ccc(cc1)N=C=S

InChI

1S/C8H4F3NOS/c9-8(10,11)13-7-3-1-6(2-4-7)12-5-14/h1-4H

InChI key

JKSZUQPHKOPVHF-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

225.0 °F - closed cup

Flash Point(C)

107.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fazila Rizvi et al.
Scientific reports, 9(1), 6738-6738 (2019-05-03)
A library of thiosemicarbazide derivatives of isoniazid 3-27, was synthesized and evaluated for their anti-inflammatory and urease inhibition activities, by using in vitro bioassays. Among these compounds 9, 10, 12, 21, and 26 were identified as new derivatives. Prolonged use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service