Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

523496

Sigma-Aldrich

4-Iodobenzyl alcohol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4CH2OH
CAS Number:
Molecular Weight:
234.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

72-75 °C (lit.)

functional group

hydroxyl
iodo

SMILES string

OCc1ccc(I)cc1

InChI

1S/C7H7IO/c8-7-3-1-6(5-9)2-4-7/h1-4,9H,5H2

InChI key

CNQRHSZYVFYOIE-UHFFFAOYSA-N

General description

4-Iodobenzyl alcohol is a benzyl alcohol derivative.

Application

4-Iodobenzyl alcohol may be used in the preparation of:
  • S-(4-Iodobenzyl) thioacetate
  • S-(4-iodobenzyl) thiobenzoate
  • fatty acid 4-iodobenzyl esters (FAIBEs)
  • (4-trimethylsilylethynylphenyl)methanol

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ferrocenyl Oligo (phenylene-vinylene) Thiols for the Construction of Self-Assembled Monolayers.
Amatore C, et al.
European Journal of Organic Chemistry, 25, 4035-4042 (2007)
Huong T Pham et al.
Rapid communications in mass spectrometry : RCM, 27(7), 805-815 (2013-03-16)
Both traditional electron ionization and electrospray ionization tandem mass spectrometry have demonstrated limitations in the unambiguous identification of fatty acids. In the former case, high electron energies lead to extensive dissociation of the radical cations from which little specific structural
Synthesis of a triblock copolymer containing a diacetylene group and its use for preparation of carbon nanodots.
Kim BJ, et al.
Macromolecular Research, 16(2), 103-107 (2008)
Daley R.
Organic Chemistry, 513-513 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service