Skip to Content
MilliporeSigma
All Photos(4)

Documents

520810

Sigma-Aldrich

Palladium black

greener alternative

99.95% trace metals basis

Synonym(s):

Precipitated palladium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pd
CAS Number:
Molecular Weight:
106.42
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

99.95% trace metals basis

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

40-60 m2/g

greener alternative category

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Palladium black (99.95% trace metal basis) can be used as a catalyst with a high surface area of ~47 m2g−1 in a variety of sustainable applications. It can be used as an anode catalyst that shows good performance and can be used in polymer exchange membrane (PEM) based fuel cells. It can also be used as an alternative catalyst to carbon-supported palladium for the hydrogenolysis reactions.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium black as model catalyst in the hydrogenolysis of CCl2F2 (CFC-12) into CH2F2 (HFC-32)
Van de Sandt E, et al.
Applied Catalysis A: General, 155(1), 59-73 (1997)
Freestanding palladium nanosheets with plasmonic and catalytic properties
Huang X, et al.
Nature Nanotechnology, 6(1), 28-28 (2011)
The behavior of palladium catalysts in direct formic acid fuel cells
Zhu Y, et al.
Journal of Power Sources, 139(1-2), 15-20 (2005)
Stefano Nicolai et al.
The Journal of organic chemistry, 78(8), 3783-3801 (2013-03-21)
Tetrahydrofurans and pyrrolidines are among the most important heterocycles found in bioactive compounds. Cyclization-functionalization domino reactions of alcohols or amines onto olefins constitute one of the most efficient methods to access them. In this context, oxy- and aminoalkynylation are especially
Meng Wang et al.
Organic & biomolecular chemistry, 11(16), 2574-2577 (2013-03-15)
A novel Pd/Cu catalyzed domino reaction for the synthesis of functionalized pyrrolo[1,2-b]pyridazines from readily accessible (hetero)aryl propargyl alcohols and 1-amino-2-bromopyrroles was developed. This cascade process involves a Sonogashira cross-coupling reaction, an isomerization and an intramolecular condensation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service