Skip to Content
MilliporeSigma
All Photos(1)

Documents

496502

Sigma-Aldrich

Tris[4-(vinyloxy)butyl] trimellitate

Synonym(s):

VEctomer® 5015 vinyl ether, Tris[4-(vinyloxy)butyl]-1,2,4-benzenetricarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3[CO2(CH2)4OCH=CH2]3
CAS Number:
Molecular Weight:
504.57
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

refractive index

n20/D 1.51 (lit.)

density

1.12 g/mL at 25 °C (lit.)

SMILES string

C=COCCCCOC(=O)c1ccc(C(=O)OCCCCOC=C)c(c1)C(=O)OCCCCOC=C

InChI

1S/C27H36O9/c1-4-31-15-7-10-18-34-25(28)22-13-14-23(26(29)35-19-11-8-16-32-5-2)24(21-22)27(30)36-20-12-9-17-33-6-3/h4-6,13-14,21H,1-3,7-12,15-20H2

InChI key

XOTMHFNWERTCLG-UHFFFAOYSA-N

Legal Information

Product of Morflex, Inc.
VEctomer is a registered trademark of Vertellus Specialties, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service