Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

496243

Sigma-Aldrich

2-Methylbenzyl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH2NCO
CAS Number:
Molecular Weight:
147.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.528 (lit.)

bp

226 °C (lit.)

density

1.059 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1ccccc1CN=C=O

InChI

1S/C9H9NO/c1-8-4-2-3-5-9(8)6-10-7-11/h2-5H,6H2,1H3

InChI key

VNHPWTGETWKSLP-UHFFFAOYSA-N

General description

2-Methylbenzyl isocyanate, also known as 1-(isocyanatomethyl)-2-methylbenzene, is an organic building block containing an isocyanate group. Its vapor pressure has been reported.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL
The Yaws Handbook of Vapor Pressure: Antoine coefficients , 77-77 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service