Skip to Content
MilliporeSigma
All Photos(1)

Documents

492183

Sigma-Aldrich

Dichloromethane-13C

99 atom % 13C

Synonym(s):

Methylene-13C chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH2Cl2
CAS Number:
Molecular Weight:
85.93
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Assay

99% (CP)

refractive index

n20/D 1.424 (lit.)

bp

40 °C (lit.)

mp

−97 °C (lit.)

density

1.341 g/mL at 25 °C

application(s)

environmental

mass shift

M+1

SMILES string

Cl[13CH2]Cl

InChI

1S/CH2Cl2/c2-1-3/h1H2/i1+1

InChI key

YMWUJEATGCHHMB-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service