Skip to Content
MilliporeSigma
All Photos(1)

Documents

481750

Sigma-Aldrich

Acetonitrile-d3

99.95 atom % D

Synonym(s):

Methyl-d3 cyanide, Trideuteroacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CN
CAS Number:
Molecular Weight:
44.07
Beilstein:
1740230
MDL number:
UNSPSC Code:
12142201

isotopic purity

99.95 atom % D

contains

1% TMS

refractive index

n20/D 1.341 (lit.)

bp

80.7 °C (lit.)

density

0.844 g/mL at 25 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])C#N

InChI

1S/C2H3N/c1-2-3/h1H3/i1D3

InChI key

WEVYAHXRMPXWCK-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service