Skip to Content
MilliporeSigma
All Photos(1)

Documents

466913

Sigma-Aldrich

2-(3-Chloropropoxy)tetrahydro-2H-pyran

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15ClO2
CAS Number:
Molecular Weight:
178.66
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.458 (lit.)

bp

55 °C/0.5 mmHg (lit.)

density

1.08 g/mL at 25 °C (lit.)

SMILES string

ClCCCOC1CCCCO1

InChI

1S/C8H15ClO2/c9-5-3-7-11-8-4-1-2-6-10-8/h8H,1-7H2

InChI key

QFXXARKSLAKVRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-(3-Chloropropoxy)tetrahydro-2H-pyran may be used in the preparation of 3,6-dihydro-6-(3-((tetrahydro-2H-pyran-2-yl)oxy)-propyl)-2(1H)-pyridinone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-Benzazepines. 9. Synthesis and chemistry of 3H-2-benzazepine and pyrimido [4, 5-d][2] benzazepine derivatives.
Trybulski EJ, et al.
The Journal of Organic Chemistry, 51(12), 2191-2202 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service