Skip to Content
MilliporeSigma
All Photos(1)

Documents

465992

Sigma-Aldrich

Magnesium

dendritic pieces, purified by distillation, 99.99% trace metals basis

Synonym(s):

Hypermagnesemia, Magnesium 100, Magnesium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
Beilstein:
4948473
EC Number:
MDL number:
UNSPSC Code:
12141502
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 621 °C)

Assay

99.99% trace metals basis

form

dendritic pieces

autoignition temp.

950 °F

purified by

distillation

reaction suitability

reagent type: reductant

resistivity

4.46 μΩ-cm, 20°C

bp

1090 °C (lit.)

mp

648 °C (lit.)

density

1.74 g/mL at 25 °C (lit.)

SMILES string

[Mg]

InChI

1S/Mg

InChI key

FYYHWMGAXLPEAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Solid state synthesis with Ca and Sn resulted in a new phase, Ca6.2Mg3.8Sn7, which has an unprecedented type of tin chain composed of square-planar tin units.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Self-heat. 1 - Water-react 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ganguli, A.K. et al.
Journal of the American Chemical Society, 120, 1223-1223 (1998)
Shuvendu Biswas et al.
Nature chemistry, 5(7), 613-620 (2013-06-22)
The development of nanocarriers that selectively release guest molecules on sensing a particular biological signal is being actively pursued in nanomedicine for diagnostic and therapeutic purposes. Here we report a protein-based nanocarrier that opens in the presence of intracellular adenosine-5'-triphosphate
Benjamin Chaigne-Delalande et al.
Science (New York, N.Y.), 341(6142), 186-191 (2013-07-13)
The magnesium transporter 1 (MAGT1) is a critical regulator of basal intracellular free magnesium (Mg(2+)) concentrations. Individuals with genetic deficiencies in MAGT1 have high levels of Epstein-Barr virus (EBV) and a predisposition to lymphoma. We show that decreased intracellular free
Carlos Zarate et al.
Annals of the New York Academy of Sciences, 1292, 21-31 (2013-07-24)
Clinical depression is a serious mental disorder characterized by low mood, anhedonia, loss of interest in daily activities, and other symptoms, and is associated with severe consequences including suicide and increased risk of cardiovascular events. Depression affects nearly 15% of
Olof Allnér et al.
RNA (New York, N.Y.), 19(7), 916-926 (2013-05-30)
Riboswitches are mRNA-based molecules capable of controlling the expression of genes. They undergo conformational changes upon ligand binding, and as a result, they inhibit or promote the expression of the associated gene. The close connection between structural rearrangement and function

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service