Skip to Content
MilliporeSigma
All Photos(1)

Documents

456519

Sigma-Aldrich

3-Cyclopentenecarboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H7CO2H
CAS Number:
Molecular Weight:
112.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.469 (lit.)

bp

215 °C (lit.)

density

1.084 g/mL at 25 °C (lit.)

SMILES string

OC(=O)C1CC=CC1

InChI

1S/C6H8O2/c7-6(8)5-3-1-2-4-5/h1-2,5H,3-4H2,(H,7,8)

InChI key

XVSYDLITVYBCBD-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

High-Field 1H and 13C NMR Spectroscopic Study of the 2-Norbornyl Cation1a.
Olah GA, et al.
Journal of the American Chemical Society, 104(25), 7105-7108 (1982)
Changxia Shi et al.
Science advances, 6(34), eabc0495-eabc0495 (2020-09-03)
Three types of seemingly unyielding trade-offs have continued to challenge the rational design for circular polymers with both high chemical recyclability and high-performance properties: depolymerizability/performance, crystallinity/ductility, and stereo-disorder/crystallinity. Here, we introduce a monomer design strategy based on a bridged bicyclic
Improved selectivity in the preparation of some 1, 1-difunctionalized 3-cyclopentenes. High yield synthesis of 3-cyclopentenecarboxylic acid.
Depres JP and Greene AE.
The Journal of Organic Chemistry, 49(5), 928-931 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service