Skip to Content
MilliporeSigma
All Photos(1)

Documents

447617

Sigma-Aldrich

Hexaethyldigermanium(IV)

97%

Synonym(s):

1,1,1,2,2,2-Hexaethyldigermane, Hexaethyldigermane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3GeGe(C2H5)3
CAS Number:
Molecular Weight:
319.65
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

reaction suitability

core: germanium

refractive index

n20/D 1.498 (lit.)

bp

219 °C (lit.)

density

1.142 g/mL at 25 °C (lit.)

SMILES string

CC[Ge](CC)(CC)[Ge](CC)(CC)CC

InChI

1S/C12H30Ge2/c1-7-13(8-2,9-3)14(10-4,11-5)12-6/h7-12H2,1-6H3

InChI key

KAZJJJFSTXNXEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service