Skip to Content
MilliporeSigma
All Photos(1)

Documents

445371

Sigma-Aldrich

(R)-(+)-α,α-Di(2-naphthyl)-2-pyrrolidinemethanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H23NO
CAS Number:
Molecular Weight:
353.46
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

99%

optical activity

[α]20/D +101°, c = 0.7 in methanol

mp

135-138 °C (lit.)

SMILES string

OC([C@H]1CCCN1)(c2ccc3ccccc3c2)c4ccc5ccccc5c4


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service