Skip to Content
MilliporeSigma
All Photos(1)

Documents

44065

Sigma-Aldrich

1,12-Dodecanediylbis(tributylphosphonium) dibromide

≥95.0% (AT)

Synonym(s):

Dodecamethylenebis(tributylphosphonium) dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H78Br2P2
CAS Number:
Molecular Weight:
732.76
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

≥95.0% (AT)

SMILES string

[Br-].[Br-].CCCC[P+](CCCC)(CCCC)CCCCCCCCCCCC[P+](CCCC)(CCCC)CCCC

Other Notes

Reagent for ion-pair chromatography

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A. Ohki et al.
Chromatographia, 32, 73-73 (1991)
Clarissa K L Ng et al.
Journal of medicinal chemistry, 49(2), 811-816 (2006-01-20)
A series of bisquaternary ammonium salts with a 12-carbon spacer between the positive charges were synthesized, and their antifungal activity has been investigated. Compounds with butyl, pentyl, and isopentyl headgroups were the most potent antifungal agents with MICs in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service