Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

434930

Sigma-Aldrich

Polyacrylamide solution

average Mw 1,500, 50 wt. % in H2O

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

mol wt

average Mw 1,500

concentration

50 wt. % in H2O

refractive index

n20/D 1.452

pH

4-6

viscosity

2-3 cP, 15 % in H2O

density

1.11 g/mL at 25 °C

SMILES string

NC(=O)C=C

InChI

1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)

InChI key

HRPVXLWXLXDGHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Binding agent, dispersing aid, lubricant, drag reduction and crystal formation control.

Features and Benefits

Tolerance for electrolytes, high initial tack and stable over a wide pH range.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyuck Joon Kwon et al.
Journal of the mechanical behavior of biomedical materials, 17, 337-346 (2012-11-07)
Many studies have demonstrated that sulfur-containing acidic groups induce chondrogenesis in vitro and in vivo. Recently, it is increasingly clear that mechanical properties of cell substrates largely influence cell differentiation. Thus, the present study investigated how mechanical properties of sulfonate-coated
Dammika P Manage et al.
Electrophoresis, 33(21), 3213-3221 (2012-10-03)
Electrophoresis is a powerful method that has seen a wide range of applications, often in automated genetic diagnostic instruments that require the use of a replaceable sieving matrix. The power and simplicity of electrophoresis as an analysis technique would be
Gennadiy Patlazhan et al.
Aesthetic plastic surgery, 37(2), 312-320 (2013-02-06)
An algorithm for breast reconstruction for patients with complications after polyacrylamide gel (PAAG) injections is presented, and the outcomes of surgery using the adopted treatment protocol are evaluated. We reviewed the clinical records of consecutive patients who were treated for
Robert E Birdsall et al.
Electrophoresis, 34(5), 753-760 (2012-12-12)
Sieving of proteins in silica colloidal crystals of millimeter dimensions is characterized for particle diameters of nominally 350 and 500 nm, where the colloidal crystals are chemically modified with a brush layer of polyacrylamide. A model is developed that relates
Honghao Sun et al.
Bioconjugate chemistry, 23(11), 2247-2255 (2012-11-02)
Our ability to design receptor-targeted nanocarriers aimed at drug release after endocytosis is limited by the current knowledge of intracellular nanoparticle (NP) trafficking. It is not clear if NP size, surface chemistry, and/or targeting of cell surface receptors changes the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service