424021
3,5,5-Trimethylhexyl acrylate
technical grade
Synonym(s):
3,5,5-Trimethylhexyl 2-propenoate, 3,5,5-Trimethylhexyl prop-2-enoate, TMHA
About This Item
Recommended Products
grade
technical grade
form
liquid
contains
20-120 ppm MEHQ as inhibitor
refractive index
n20/D 1.437 (lit.)
bp
55 °C/0.4 mmHg (lit.)
mp
−34 °C (lit.)
density
0.875 g/mL at 25 °C (lit.)
SMILES string
CC(CCOC(=O)C=C)CC(C)(C)C
InChI
1S/C12H22O2/c1-6-11(13)14-8-7-10(2)9-12(3,4)5/h6,10H,1,7-9H2,2-5H3
InChI key
FIXKCCRANLATRP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
192.2 °F - closed cup
Flash Point(C)
89 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service