Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

415103

Sigma-Aldrich

Calcium nitride

−200 mesh, 95%

Synonym(s):

Tricalcium dinitride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca3N2
CAS Number:
Molecular Weight:
148.25
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

powder

particle size

−200 mesh

density

2.63 g/mL at 25 °C (lit.)

SMILES string

[Ca]=N[Ca]N=[Ca]

InChI

1S/3Ca.2N

InChI key

DVJMVZQXJNJSJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service