Skip to Content
MilliporeSigma
All Photos(1)

Documents

413402

Sigma-Aldrich

Iron(II) acetylacetonate

99.95% trace metals basis

Synonym(s):

2,4-Pentanedione iron(II) derivative, Fe(acac)2, Ferrous acetylacetonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3COCH=C(O)CH3]2Fe
CAS Number:
Molecular Weight:
254.06
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.95% trace metals basis

form

solid

reaction suitability

core: iron

mp

175 °C (dec.) (lit.)

SMILES string

CC(=O)\C=C(\C)O[Fe]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Fe/c2*1-4(6)3-5(2)7;/h2*3,6H,1-2H3;/q;;+2/p-2/b2*4-3-;

InChI key

LFORAFQNBQKDRY-FDGPNNRMSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 26 Iron

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Li et al.
Nanoscale, 12(19), 10834-10841 (2020-05-13)
ε-Iron nitrides with the general formula ε-Fe3N1+x (-0.40 < x < 0.48) have been widely studied due to their interesting magnetism. However, the phase diagram of the Fe-N binary system indicates the absence of monophasic ε-Fe3N1+x (x < 0) compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service