Skip to Content
MilliporeSigma
All Photos(1)

Documents

409391

Sigma-Aldrich

Benzyl(chloromethyl)dimethylsilane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2Si(CH3)2CH2Cl
CAS Number:
Molecular Weight:
198.76
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.519 (lit.)

bp

120-124 °C/40 mmHg (lit.)

density

1.01 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(CCl)Cc1ccccc1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service