Skip to Content
MilliporeSigma
All Photos(1)

Documents

398357

Sigma-Aldrich

Hydrogen hexabromoplatinate(IV) hydrate

99.9% trace metals basis

Synonym(s):

Bromoplatinic acid, Platinic bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2PtBr6·xH2O
CAS Number:
Molecular Weight:
676.52 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99.9% trace metals basis

form

powder and chunks

composition

Pt, 24-26%

SMILES string

O.Br.Br.Br[Pt](Br)(Br)Br

InChI

1S/6BrH.H2O.Pt/h6*1H;1H2;/q;;;;;;;+4/p-4

InChI key

YTNOPOJYUCYUMR-UHFFFAOYSA-J

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service