Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

380342

Sigma-Aldrich

(1S,2S,5S)-(−)-2-Hydroxy-3-pinanone

99%

Synonym(s):

(1S,2S,5S)-2-Hydroxy-2,6,6-trimethylbicyclo[3.1.1]heptan-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O2
CAS Number:
Molecular Weight:
168.23
Beilstein:
3196635
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

optical activity

[α]20/D −39°, c = 0.5 in chloroform

bp

245 °C (lit.)

mp

36-38 °C (lit.)

density

1.059 g/mL at 25 °C (lit.)

SMILES string

CC1(C)[C@H]2C[C@@H]1[C@](C)(O)C(=O)C2

InChI

1S/C10H16O2/c1-9(2)6-4-7(9)10(3,12)8(11)5-6/h6-7,12H,4-5H2,1-3H3/t6-,7-,10-/m0/s1

InChI key

VZRRCQOUNSHSGB-BYULHYEWSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service